Order Entry
Germany
Orders LinkContactUsLinkComponent
1-Octadecanol, Sigma-Aldrich®

1-Octadecanol, Sigma-Aldrich®

Plastic bottle for solids, 100 g
Catalog # SIAL258768-100G
Supplier:   Merck
1-Octadecanol, Sigma-Aldrich®
Catalog # SIAL258768-100G
Supplier:  Merck
CAS Number:  112-92-5
1-Octadecanol, Sigma-Aldrich®

Plastic bottle for solids, 100 g

CAS Number:  112-92-5
Price
80,10 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
  • CAS Number:112-92-5
  • Formula:H₃C(CH₂)₁₆CH₂OH
  • MDL Number:MFCD00002823
  • Molecular Weight:270.5 g/mol
  • Merck Index:12,08960
  • Boiling Pt:336 °C (1013 hPa)
  • Melting Pt:56…57 °C
  • Density:0.805…0.815 g/cm³ (60 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:204-017-6
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.