Order Entry
Germany
Orders LinkContactUsLinkComponent
Decontamination Solution, DNA/RNA-ExitusPlus™

Decontamination Solution, DNA/RNA-ExitusPlus™

DNA/RNA-ExitusPlus™, spray bottle, 100 mL
Catalog # A7089.0100
Supplier:   PanReac AppliChem
Decontamination Solution, DNA/RNA-ExitusPlus™
Catalog # A7089.0100
Supplier:  PanReac AppliChem
CAS Number:  
Decontamination Solution, DNA/RNA-ExitusPlus™
Decontamination Solution, DNA/RNA-ExitusPlus™

DNA/RNA-ExitusPlus™, spray bottle, 100 mL

CAS Number:  
Price
43,57 €
Log in for availability
1 * 100 mL
Product Details & Documents

Solution for the removal of DNA and RNA contamination from surfaces. Suitable for cleaning PCR work stations and equipment, electrophoresis equipment, as well as pipettes and reaction tubes, for example.

  • All components are readily bio-degradable and not harmful or toxic for humans
  • Doesn't contain aggressive mineral acids or alkaline substances
  • No organic solvents or volatile components
  • Delivered as spray bottles or refill bottles (RF)
Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    DNA/RNA-ExitusPlus™, spray bottle
  • Cat. No.:
    A7089.0100
  • Storage temperature:
    Ambient

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.