Orders LinkContactUsLinkComponent
Soil samplers, Mole
Soil samplers, Mole
Drill bit for sand
Catalog # BURK5350-1020
Supplier:   BURKLE
Soil samplers, Mole
Catalog # BURK5350-1020
Supplier:  BURKLE
Soil samplers, Mole
Soil samplers, Mole
Drill bit for sand
Price
311,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Very robust model with specially hardened forged drill bits. For determining soil properties, moisture and structure. The shape of the Mole‘s drill bits ensures minimum friction when drilling and removing the Mole from the ground.The Mole has different components. Handle with 75 cm rod, extension rod 100 cm if necessary and one of seven available drill bits, each of which is designed for a particular soil type. The components can be combined without any problems. Extension rods can extend the working depth of the Mole down to 5 m. Please order drill bits seperately.
  • Physical efforts are reduced to a minimum
Specifications
  • Description:
    Drill bit for sand
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Extension rod (100 cm)
    Extension rod (100 cm)
    BURK5350-1010
    Log in for availability
    1 * 1 ST - 175,00 €