1M LiCl non-aqueous filling solution in ethanol 1 * 100 mL
Catalog # REGEEFSLIET
Lieferant: REAGECON
1M LiCl non-aqueous filling solution in ethanol 1 * 100 mL
Catalog # REGEEFSLIET
Lieferant: REAGECON
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Lieferant: VWR Chemicals
Acetone ≥99%, TECHNICAL
Lieferant: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Lieferant: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Lieferant: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Lieferant: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Lieferant: Merck
VWR® Aquaresist Water Bath Protective Agent
Lieferant: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Lieferant: Dr. Weigert
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Lieferant: VWR Chemicals
VWR®, Nitrile Gloves
Lieferant: VWR Collection
VWR® Facial Tissues
Lieferant: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Lieferant: KIMBERLY CLARK
pH indicator strips, MQuant®, Supelco®
Lieferant: Merck