Orders LinkContactUsLinkComponent
Clamps
Clamps
Flat flanges DN 100
Catalog # LENZ06247010
Supplier:   Lenz Laborglas
Clamps
Catalog # LENZ06247010
Supplier:  Lenz Laborglas
Clamps
Clamps
Flat flanges DN 100
Price
1 440,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Excellent chemical resistance
  • Excellent temperature resistance
For fixing lids onto reaction vessels, available for flat flanges LF100-200.
Specifications
  • For:
    Flat flanges DN 100
  • Seq:
    01