Orders LinkContactUsLinkComponent
Petri dishes, disposable | Avantor
Petri dishes, disposable
With 2 compartments, 90 mm, Sterile
Catalog # KART362
Supplier:   KARTELL
Petri dishes, disposable
Catalog # KART362
Supplier:  KARTELL
Petri dishes, disposable
Petri dishes, disposable
With 2 compartments, 90 mm, Sterile
Wechseln zu:
Price
421,00 €
Anmelden für die Verfügbarkeit
1 * 500 ST
Product Details & Documents
Disposable planar, clear Petri dishes in PS. Available in several diameters, ventilated and unventilated, aseptic or radiation-sterilised. Styles with 2, 3 and 4 separate compartments. Designed for routine lab work and bacteriological research. Available with aeration notches on the lid to prevent condensation formation, or without aeration notches to avoid agar dessication after incubation.
Spezifikationen:
  • Ø:
    90 mm
  • Sterility:
    Sterile
  • Version:
    With 2 compartments