Orders LinkContactUsLinkComponent
Petri dishes, disposable
Petri dishes, disposable
Without vents, 60 mm, Sterile
Catalog # KART360
Supplier:   KARTELL
Petri dishes, disposable
Catalog # KART360
Supplier:  KARTELL
Petri dishes, disposable
Petri dishes, disposable
Without vents, 60 mm, Sterile
Price
287,00 €
Log in for availability
1 * 500 ST
Product Details & Documents
Disposable planar, clear Petri dishes in PS. Available in several diameters, ventilated and unventilated, aseptic or radiation-sterilised. Styles with 2, 3 and 4 separate compartments. Designed for routine lab work and bacteriological research. Available with aeration notches on the lid to prevent condensation formation, or without aeration notches to avoid agar dessication after incubation.
Specifications
  • Ø:
    60 mm
  • Sterility:
    Sterile
  • Version:
    Without vents