Order Entry
Germany
Orders LinkContactUsLinkComponent
Flat spatula, conical

Flat spatula, conical

Nickel, 180 mm, 20/35 mm
Catalog # 231-0126
Supplier:   NICKEL ELECTRO
Flat spatula, conical
Catalog # 231-0126
Supplier:  NICKEL ELECTRO
Flat spatula, conical
Flat spatula, conical

Nickel, 180 mm, 20/35 mm

Price
71,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel or pure nickel (99,6%). Multipurpose spatula for a wide range of uses.

  • Excellent chemical resistance
  • Excellent thermal resistance
  • Smooth surfaces for fast, easy cleaning
Specifications
  • Description:
    Nickel
  • Length:
    180 mm
  • Blade width:
    20/35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.