Orders LinkContactUsLinkComponent
Adapters, PTFE, socket on cone
Adapters, PTFE, socket on cone
full length, A, 24/40 NS, 19/38 NS
Catalog # COWI021.01924
Supplier:   COWIE
Adapters, PTFE, socket on cone
Catalog # COWI021.01924
Supplier:  COWIE
Adapters, PTFE, socket on cone
Adapters, PTFE, socket on cone
full length, A, 24/40 NS, 19/38 NS
Price
24,70 €
Log in for availability
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Made of pure PTFE, white.
  • Very good chemical resistance
  • Very good temperature resistance
  • Fully adjustable with replacement Viton® seal
  • Contact materials are glass and PTFE only
This holder enables standard probes and thermometers to be assembled in standard taper laboratory glassware.
Specifications
  • Cone size:
    24/40 NS
  • Socket size:
    19/38 NS
  • Type:
    full length, A
  • Seq:
    05