Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Ethoxybenzonitrile ≥98%

4-Ethoxybenzonitrile ≥98%

Glass bottle for solids, 100 g
Catalog # A19806.22
Toxic
Irritant
4-Ethoxybenzonitrile ≥98%
Catalog # A19806.22
CAS Number:  25117-74-2
4-Ethoxybenzonitrile ≥98%
4-Ethoxybenzonitrile ≥98%

Glass bottle for solids, 100 g

CAS Number:  25117-74-2
Price
481,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 25117-74-2 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 60-63° Boiling Point: 258° UN No.: 3439 MDL No.: MFCD00001819

Synonyms:  p-Ethoxybenzonitrile
  • CAS Number:25117-74-2
  • Formula:C₉H₉NO
  • MDL Number:MFCD00001819
  • Molecular Weight:147.18 g/mol
  • Boiling Pt:258 °C
  • Melting Pt:60…63 °C
  • Storage Temperature:Ambient
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.