Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Ethoxybenzonitrile ≥98%

4-Ethoxybenzonitrile ≥98%

Glass bottle for solids, 5 g
Catalog # A19806.06
Toxic
Irritant
4-Ethoxybenzonitrile ≥98%
Catalog # A19806.06
CAS Number:  25117-74-2
4-Ethoxybenzonitrile ≥98%
4-Ethoxybenzonitrile ≥98%

Glass bottle for solids, 5 g

CAS Number:  25117-74-2
Price
40,30 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 25117-74-2 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 60-63° Boiling Point: 258° UN No.: 3439 MDL No.: MFCD00001819

Synonyms:  p-Ethoxybenzonitrile
  • CAS Number:25117-74-2
  • Formula:C₉H₉NO
  • MDL Number:MFCD00001819
  • Molecular Weight:147.18 g/mol
  • Boiling Pt:258 °C
  • Melting Pt:60…63 °C
  • Storage Temperature:Ambient
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications