Order Entry
Germany
Orders LinkContactUsLinkComponent
Anthrone 96%

Anthrone 96%

Glass bottle for solids, 500 g
Catalog # A19118.36
Health
Irritant
Anthrone 96%
Catalog # A19118.36
CAS Number:  90-44-8
Anthrone 96%
Anthrone 96%

Glass bottle for solids, 500 g

CAS Number:  90-44-8
Price
624,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 97% CAS No.: 90-44-8 Molecular Formula: C14H10O Formula Weight: 194.23 Melting Point: 153-160° Solubility: Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water Beilstein Reference: 1910173 Merck Reference: 14,691 MDL No.: MFCD00001187

In organic synthesis, in Colourimetric determination of sugar in body fluids and of animal starch in liver tissue

Synonyms:  Anthracen-9(10H)-one
  • CAS Number:90-44-8
  • Formula:C₁₄H₁₀O
  • MDL Number:MFCD00001187
  • Molecular Weight:194.23 g/mol
  • Merck Index:14,691
  • Melting Pt:153…160 °C
  • Storage Temperature:Ambient
  • EINECS:201-994-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.