Order Entry
Germany
Orders LinkContactUsLinkComponent
Anthrone 96%

Anthrone 96%

Glass bottle for solids, 100 g
Catalog # A19118.22
Health
Irritant
Anthrone 96%
Catalog # A19118.22
CAS Number:  90-44-8
Anthrone 96%
Anthrone 96%

Glass bottle for solids, 100 g

CAS Number:  90-44-8
Price
180,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 97% CAS No.: 90-44-8 Molecular Formula: C14H10O Formula Weight: 194.23 Melting Point: 153-160° Solubility: Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water Beilstein Reference: 1910173 Merck Reference: 14,691 MDL No.: MFCD00001187

In organic synthesis, in Colourimetric determination of sugar in body fluids and of animal starch in liver tissue

Synonyms:  Anthracen-9(10H)-one
  • CAS Number:90-44-8
  • Formula:C₁₄H₁₀O
  • MDL Number:MFCD00001187
  • Molecular Weight:194.23 g/mol
  • Merck Index:14,691
  • Melting Pt:153…160 °C
  • Storage Temperature:Ambient
  • EINECS:201-994-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications