Sodium nitrite ≥98%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 7632-00-0 Molecular Formula: NNaO2 Formula Weight: 69.00 Odour: Odourless Melting Point: 271° dec. UN No.: 1500 MDL No.: MFCD00011118
Synonyms: Diazotizing salts
- CAS Number:7632-00-0
- Formula:NaNO₂
- MDL Number:MFCD00011118
- Molecular Weight:69 g/mol
- Merck Index:14,8648
- Boiling Pt:320 °C (1013 hPa)
- Melting Pt:271 °C (dec.)
- Density:2.17
- Storage Temperature:Ambient
- EINECS:231-555-9
- UN:1500
- ADR:5.1,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



