Order Entry
Germany
Orders LinkContactUsLinkComponent
n-Nonane ≥99%

n-Nonane ≥99%

Glass bottle, 100 mL
Catalog # A16177.AE
Health
Flammable
Irritant
n-Nonane ≥99%
Catalog # A16177.AE
CAS Number:  111-84-2
n-Nonane ≥99%
n-Nonane ≥99%

Glass bottle, 100 mL

CAS Number:  111-84-2
Price
41,60 €
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 99% CAS No.: 111-84-2 Molecular Formula: C9H20 Formula Weight: 128.26 Flash Point: 31°(87°F) Refractive Index: 1.4054 Melting Point: -53° Boiling Point: 150-152° Beilstein Reference: 1696917 UN No.: 1920 MDL No.: MFCD00009574

  • CAS Number:111-84-2
  • Formula:H₃C(CH₂)₇CH₃
  • MDL Number:MFCD00009574
  • Molecular Weight:128.26 g/mol
  • Boiling Pt:150…152 °C
  • Melting Pt:–53 °C
  • Density:0.718
  • Flash Pt:31 °C (87 °F)
  • Storage Temperature:Ambient
  • EINECS:203-913-4
  • UN:1920
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications