Order Entry
Germany
Orders LinkContactUsLinkComponent
PIPES ≥98%

PIPES ≥98%

Glass bottle for solids, 500 g
Catalog # A16090.36
PIPES ≥98%
Catalog # A16090.36
CAS Number:  5625-37-6
PIPES ≥98%
PIPES ≥98%

Glass bottle for solids, 500 g

CAS Number:  5625-37-6
Price
407,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98% CAS No.: 5625-37-6 Molecular Formula: C8H18N2O6S2 Formula Weight: 302.37 Melting Point: 300° Beilstein Reference: 817713 Merck Reference: 14,7479 MDL No.: MFCD00006159

Synonyms:  Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid
  • CAS Number:5625-37-6
  • Formula:C₈H₁₈N₂O₆S₂
  • MDL Number:MFCD00006159
  • Molecular Weight:302.37 g/mol
  • Merck Index:14,7479
  • Boiling Pt:499 °C (1013 hPa)
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:227-057-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications