Order Entry
Germany
Orders LinkContactUsLinkComponent
 

PIPES ≥98%

Glass bottle for solids, 100 g
Catalog # A16090.22
PIPES ≥98%
Catalog # A16090.22
CAS Number:  5625-37-6
PIPES ≥98%
PIPES ≥98%

Glass bottle for solids, 100 g

CAS Number:  5625-37-6
Price
99,60 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 5625-37-6 Molecular Formula: C8H18N2O6S2 Formula Weight: 302.37 Melting Point: 300° Beilstein Reference: 817713 Merck Reference: 14,7479 MDL No.: MFCD00006159

Synonyms:  Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid
  • CAS Number:5625-37-6
  • Formula:C₈H₁₈N₂O₆S₂
  • MDL Number:MFCD00006159
  • Molecular Weight:302.37 g/mol
  • Merck Index:14,7479
  • Boiling Pt:499 °C (1013 hPa)
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:227-057-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.