Order Entry
Germany
Orders LinkContactUsLinkComponent
Methylcyclohexane ≥99%

Methylcyclohexane ≥99%

Glass bottle, 2.500 mL
Catalog # A16057.0F
Flammable
Health
Irritant
Environment
Methylcyclohexane ≥99%
Catalog # A16057.0F
CAS Number:  108-87-2
Methylcyclohexane ≥99%
Methylcyclohexane ≥99%

Glass bottle, 2.500 mL

CAS Number:  108-87-2
Price
78,50 €
Log in for availability
1 * 2.500 mL
Product Details & Documents
  • CAS Number:108-87-2
  • Formula:C₆H₁₁CH₃
  • MDL Number:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:14,6047
  • Boiling Pt:101 °C
  • Melting Pt:–126 °C
  • Density:0.771
  • Flash Pt:–4 °C (–25 °F)
  • Storage Temperature:Ambient
  • EINECS:203-624-3
  • UN:2296
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.