Order Entry
Germany
Orders LinkContactUsLinkComponent
Nicotinamide ≥99%

Nicotinamide ≥99%

Glass bottle for solids, 250 g
Catalog # A15970.30
Irritant
Nicotinamide ≥99%
Catalog # A15970.30
CAS Number:  98-92-0
Nicotinamide ≥99%
Nicotinamide ≥99%

Glass bottle for solids, 250 g

CAS Number:  98-92-0
Price
54,20 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 98-92-0 Molecular Formula: C6H6N2O Formula Weight: 122.13 Flash Point: 182°(360°F) Physical Form: Powder Density: 1.40 Appearance: White Odour: Odourless Melting Point: 128-132° Beilstein Reference: 383619 Merck Reference: 14,6523 MDL No.: MFCD00006395

The amide of nicotinic acid (vitamin B-3)

Synonyms:  3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
  • CAS Number:98-92-0
  • Formula:C₆H₆N₂O
  • MDL Number:MFCD00006395
  • Molecular Weight:122.13 g/mol
  • Merck Index:14,6523
  • Boiling Pt:334 °C (1013 hPa)
  • Melting Pt:128…132 °C
  • Density:1.40
  • Flash Pt:182 °C (360 °F)
  • Storage Temperature:Ambient
  • EINECS:202-713-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications