4-Chlorophenol ≥99%
Glass bottle for solids, 1.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 106-48-9 Molecular Formula: C6H5ClO Formula Weight: 128.56 Flash Point: 115°(239°F) Odour: Phenol-like Boiling Point: 217-220° Beilstein Reference: 507004 Merck Reference: 14,2154 UN No.: 2020 MDL No.: MFCD00002318
Synonyms: p-Chlorophenol
- CAS Number:106-48-9
- Formula:C₆H₅ClO
- MDL Number:MFCD00002318
- Molecular Weight:128.56 g/mol
- Merck Index:14,2154
- Boiling Pt:217…220 °C
- Melting Pt:39…43 °C
- Density:1.306
- Flash Pt:115 °C (239 °F)
- Storage Temperature:Ambient
- EINECS:203-402-6
- UN:2020
- ADR:6.1,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1.000 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



