Chromone-3-carboxaldehyde ≥97%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Chromone-3-carboxaldehyde ≥97%
Catalog # A15438.22
Supplier: Thermo Fisher Scientific
CAS Number: 17422-74-1
Wechseln zu:
Product Details & Documents
Purity: 97% CAS No.: 17422-74-1 Molecular Formula: C10H6O3 Formula Weight: 174.16 Storage Temperature: 2°C to 8°C Melting Point: 150-154° Beilstein Reference: 472849 MDL No.: MFCD00014667
Synonyme: 4-Oxo-4H-chromene-3-carbaldehyde
- CAS-Nummer:17422-74-1
- Formel::C₁₀H₆O₃
- MDL-Nummer:MFCD00014667
- Molekulargewicht:174.16 g/mol
- Schmelzpunkt:150…154 °C
- EINECS:241-451-5
Spezifikationen:
Spezifikationen:
- Pack type:Glass bottle for solids
- Pk:100 g



