Order Entry
Germany
Orders LinkContactUsLinkComponent
Chromone-3-carboxaldehyde ≥97%

Chromone-3-carboxaldehyde ≥97%

Glass bottle for solids, 5 g
Catalog # A15438.06
Chromone-3-carboxaldehyde ≥97%
Catalog # A15438.06
CAS Number:  17422-74-1
Chromone-3-carboxaldehyde ≥97%
Chromone-3-carboxaldehyde ≥97%

Glass bottle for solids, 5 g

CAS Number:  17422-74-1
Price
34,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 17422-74-1 Molecular Formula: C10H6O3 Formula Weight: 174.16 Storage Temperature: 2°C to 8°C Melting Point: 150-154° Beilstein Reference: 472849 MDL No.: MFCD00014667

Synonyms:  4-Oxo-4H-chromene-3-carbaldehyde
  • CAS Number:17422-74-1
  • Formula:C₁₀H₆O₃
  • MDL Number:MFCD00014667
  • Molecular Weight:174.16 g/mol
  • Melting Pt:150…154 °C
  • EINECS:241-451-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.