Auftragserfassung
Germany
AuftragsverfolgungKontakt
 

Bortrifluorid-Diethylether-Komplex ≥98%

Glass bottle, 1.000 mL
Catalog # A15275.AU
Toxic
Health
Corrosive
Flammable
Bortrifluorid-Diethylether-Komplex ≥98%
Catalog # A15275.AU
CAS Number:  109-63-7
Bortrifluorid-Diethylether-Komplex ≥98%
Bortrifluorid-Diethylether-Komplex ≥98%

Glass bottle, 1.000 mL

CAS Number:  109-63-7
Price
121,00 €
Log in for availability
1 * 1.000 mL
Product Details & Documents

Purity: 98+% CAS No.: 109-63-7 Molecular Formula: C4H10BF3O Formula Weight: 141.93 Flash Point: 47°(116°F) Refractive Index: 1.3440 Odour: Acrid Melting Point: -60° Boiling Point: 125-126° Beilstein Reference: 3909607 Merck Reference: 14,1350 UN No.: 2604 MDL No.: MFCD00013194

Synonyms:  Bortrifluorid-Diethylether, Bortrifluorid-Diethyletherat, Bortrifluorid-Ethyletherat, Diethylether-Bortrifluorid
  • CAS Number:109-63-7
  • Formula:C₄H₁₀BF₃O
  • MDL Number:MFCD00013194
  • Molecular Weight:141.93 g/mol
  • Merck Index:14,1350
  • Boiling Pt:125…126 °C
  • Melting Pt:–60 °C
  • Density:1,130
  • Flash Pt:47 °C (116 °F)
  • Storage Temperature:Raumtemperatur
  • EINECS:203-689-8
  • UN:2604
  • ADR:8,I
Specifications

Specifications

  • Packung:
    Glass bottle
  • VE:
    1.000 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.