Order Entry
Germany
Orders LinkContactUsLinkComponent
Boron trifluoride diethyl ether complex ≥98%

Boron trifluoride diethyl ether complex ≥98%

Glass bottle, 100 mL
Catalog # A15275.AE
Toxic
Health
Corrosive
Flammable
Boron trifluoride diethyl ether complex ≥98%
Catalog # A15275.AE
CAS Number:  109-63-7
Boron trifluoride diethyl ether complex ≥98%
Boron trifluoride diethyl ether complex ≥98%

Glass bottle, 100 mL

CAS Number:  109-63-7
Price
33,20 €
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 98+% CAS No.: 109-63-7 Molecular Formula: C4H10BF3O Formula Weight: 141.93 Flash Point: 47°(116°F) Refractive Index: 1.3440 Odour: Acrid Melting Point: -60° Boiling Point: 125-126° Beilstein Reference: 3909607 Merck Reference: 14,1350 UN No.: 2604 MDL No.: MFCD00013194

Synonyms:  Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate
  • CAS Number:109-63-7
  • Formula:C₄H₁₀BF₃O
  • MDL Number:MFCD00013194
  • Molecular Weight:141.93 g/mol
  • Merck Index:14,1350
  • Boiling Pt:125…126 °C
  • Melting Pt:–60 °C
  • Density:1.130
  • Flash Pt:47 °C (116 °F)
  • Storage Temperature:Ambient
  • EINECS:203-689-8
  • UN:2604
  • ADR:8,I
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.