Order Entry
Germany
Orders LinkContactUsLinkComponent
3,5-Difluorophenol ≥98%

3,5-Difluorophenol ≥98%

Glass bottle for solids, 25 g
Catalog # A15177.14
Toxic
Corrosive
3,5-Difluorophenol ≥98%
Catalog # A15177.14
CAS Number:  2713-34-0
3,5-Difluorophenol ≥98%
3,5-Difluorophenol ≥98%

Glass bottle for solids, 25 g

CAS Number:  2713-34-0
Price
240,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 2713-34-0 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 70°(158°F) Odour: Phenol-like Boiling Point: 65-68°/1mm Beilstein Reference: 2078616 UN No.: 3261 MDL No.: MFCD00002255

  • CAS Number:2713-34-0
  • Formula:C₆H₄F₂O
  • MDL Number:MFCD00002255
  • Molecular Weight:130.09 g/mol
  • Boiling Pt:65…68 °C (1 mmHg)
  • Melting Pt:51…55 °C
  • Density:1.566 g/cm³ (20 °C)
  • Flash Pt:70 °C (158 °F)
  • Storage Temperature:Ambient
  • UN:3261
  • ADR:8,II
  •  :Density: 1.566 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.