Order Entry
Germany
Orders LinkContactUsLinkComponent
 

3,5-Difluorophenol ≥98%

Glass bottle for solids, 5 g
Catalog # A15177.06
Toxic
Corrosive
3,5-Difluorophenol ≥98%
Catalog # A15177.06
CAS Number:  2713-34-0
3,5-Difluorophenol ≥98%
3,5-Difluorophenol ≥98%

Glass bottle for solids, 5 g

CAS Number:  2713-34-0
Price
71,80 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98+% CAS No.: 2713-34-0 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 70°(158°F) Odour: Phenol-like Boiling Point: 65-68°/1mm Beilstein Reference: 2078616 UN No.: 3261 MDL No.: MFCD00002255

  • CAS Number:2713-34-0
  • Formula:C₆H₄F₂O
  • MDL Number:MFCD00002255
  • Molecular Weight:130.09 g/mol
  • Boiling Pt:65…68 °C (1 mmHg)
  • Melting Pt:51…55 °C
  • Density:1.566 g/cm³ (20 °C)
  • Flash Pt:70 °C (158 °F)
  • Storage Temperature:Ambient
  • UN:3261
  • ADR:8,II
  •  :Density: 1.566 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications