Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Bromothiazole ≥99%

2-Bromothiazole ≥99%

Glass bottle for solids, 50 g
Catalog # A14838.18
Irritant
2-Bromothiazole ≥99%
Catalog # A14838.18
CAS Number:  3034-53-5
2-Bromothiazole ≥99%
2-Bromothiazole ≥99%

Glass bottle for solids, 50 g

CAS Number:  3034-53-5
Price
137,00 €
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 99% CAS No.: 3034-53-5 Molecular Formula: C3H2BrNS Formula Weight: 164.02 Storage Temperature: 2°C to 8°C Flash Point: 63°(145°F) Refractive Index: 1.5930 Boiling Point: 171° Beilstein Reference: 105724 MDL No.: MFCD00005316

  • CAS Number:3034-53-5
  • Formula:C₃H₂BrNS
  • MDL Number:MFCD00005316
  • Molecular Weight:164.03 g/mol
  • Boiling Pt:171 °C
  • Density:1.836
  • Flash Pt:63 °C (145 °F)
  • EINECS:221-229-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.