Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Bromothiazole ≥99%

2-Bromothiazole ≥99%

Glass bottle for solids, 10 g
Catalog # A14838.09
Irritant
2-Bromothiazole ≥99%
Catalog # A14838.09
CAS Number:  3034-53-5
2-Bromothiazole ≥99%
2-Bromothiazole ≥99%

Glass bottle for solids, 10 g

CAS Number:  3034-53-5
Price
41,20 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 99% CAS No.: 3034-53-5 Molecular Formula: C3H2BrNS Formula Weight: 164.02 Storage Temperature: 2°C to 8°C Flash Point: 63°(145°F) Refractive Index: 1.5930 Boiling Point: 171° Beilstein Reference: 105724 MDL No.: MFCD00005316

  • CAS Number:3034-53-5
  • Formula:C₃H₂BrNS
  • MDL Number:MFCD00005316
  • Molecular Weight:164.03 g/mol
  • Boiling Pt:171 °C
  • Density:1.836
  • Flash Pt:63 °C (145 °F)
  • EINECS:221-229-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications