Order Entry
Germany
Orders LinkContactUsLinkComponent
Potassium nitrate ≥99%

Potassium nitrate ≥99%

Glass bottle for solids, 10.000 g
Catalog # A14527.0C
Oxidizing
Potassium nitrate ≥99%
Catalog # A14527.0C
CAS Number:  7757-79-1
Potassium nitrate ≥99%
Potassium nitrate ≥99%

Glass bottle for solids, 10.000 g

CAS Number:  7757-79-1
Price
260,00 €
Log in for availability
1 * 10.000 g
Product Details & Documents

Purity: 99% CAS No.: 7757-79-1 Molecular Formula: KNO3 Formula Weight: 101.11 Odour: Odourless Melting Point: 334° Boiling Point: 400° dec. Merck Reference: 14,7648 UN No.: 1486 MDL No.: MFCD00011409

Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Merck Index:14,7648
  • Boiling Pt:400 °C (dec.)
  • Melting Pt:334 °C
  • Density:2.109
  • Storage Temperature:Ambient
  • EINECS:231-818-8
  • UN:1486
  • ADR:5.1,III
  • REACH:01-2119488224-35
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10.000 g

Specifications