Order Entry
Germany
Orders LinkContactUsLinkComponent
 

2-(4-Bromophenyl)ethanol ≥98%

Glass bottle for solids, 25 g
Catalog # A14143.14
2-(4-Bromophenyl)ethanol ≥98%
Catalog # A14143.14
CAS Number:  4654-39-1
2-(4-Bromophenyl)ethanol ≥98%
2-(4-Bromophenyl)ethanol ≥98%

Glass bottle for solids, 25 g

CAS Number:  4654-39-1
Price
401,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 4654-39-1 Molecular Formula: C8H9BrO Formula Weight: 201.06 Flash Point: 110°(230°F) Refractive Index: 1.5730 Boiling Point: 106-108°/1mm Beilstein Reference: 2079929 MDL No.: MFCD00002897

Synonyms:  4-Bromophenethyl alcohol
  • CAS Number:4654-39-1
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00002897
  • Molecular Weight:201.06 g/mol
  • Boiling Pt:106…108 °C (1 mmHg)
  • Melting Pt:36…38 °C
  • Density:1.436
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:225-093-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.