Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Hydroxy-5-nitropyridine ≥98%

2-Hydroxy-5-nitropyridine ≥98%

Glass bottle for solids, 100 g
Catalog # A13921.22
Irritant
2-Hydroxy-5-nitropyridine ≥98%
Catalog # A13921.22
CAS Number:  5418-51-9
2-Hydroxy-5-nitropyridine ≥98%
2-Hydroxy-5-nitropyridine ≥98%

Glass bottle for solids, 100 g

CAS Number:  5418-51-9
Price
159,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 5418-51-9 Molecular Formula: C5H4N2O3 Formula Weight: 140.10 Melting Point: 188-191° Beilstein Reference: 120352

Synonyms:  5-Nitro-2-pyridinol
  • CAS Number:5418-51-9
  • Formula:C₅H₄N₂O₃
  • MDL Number:MFCD00006276
  • Molecular Weight:140.1 g/mol
  • Boiling Pt:180…200 °C (0.2 mmHg)
  • Melting Pt:188…191 °C
  • Storage Temperature:Ambient
  • EINECS:226-525-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications