Order Entry
Germany
Orders LinkContactUsLinkComponent
n-Eicosane ≥99%

n-Eicosane ≥99%

Glass bottle for solids, 100 g
Catalog # A13853.22
n-Eicosane ≥99%
Catalog # A13853.22
CAS Number:  112-95-8
n-Eicosane ≥99%
n-Eicosane ≥99%

Glass bottle for solids, 100 g

CAS Number:  112-95-8
Price
77,80 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 112-95-8 Molecular Formula: CH3(CH2)18CH3 Formula Weight: 282.54 Flash Point: 176°(348°F) Refractive Index: 1.4425 Melting Point: 36-38° Boiling Point: 220°/30mm Beilstein Reference: 1700722 MDL No.: MFCD00009344

Synonyms:  N-Eicosane
  • CAS Number:112-95-8
  • Formula:H₃C(CH₂)₁₈CH₃
  • MDL Number:MFCD00009344
  • Molecular Weight:282.55 g/mol
  • Boiling Pt:220 °C (30 mmHg)
  • Melting Pt:36…38 °C
  • Density:0.7888
  • Flash Pt:176 °C (348 °F)
  • Storage Temperature:Ambient
  • EINECS:204-018-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications