Order Entry
Germany
Orders LinkContactUsLinkComponent
ω,4-Dibromoacetophenone ≥98%

ω,4-Dibromoacetophenone ≥98%

Glass bottle for solids, 25 g
Catalog # A13687.14
Corrosive
ω,4-Dibromoacetophenone ≥98%
Catalog # A13687.14
CAS Number:  99-73-0
ω,4-Dibromoacetophenone ≥98%
ω,4-Dibromoacetophenone ≥98%

Glass bottle for solids, 25 g

CAS Number:  99-73-0
Price
71,50 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 99-73-0 Molecular Formula: C8H6Br2O Formula Weight: 277.95 Storage Temperature: 2°C to 8°C Odour: Pungent Beilstein Reference: 607604 Merck Reference: 14,1427 UN No.: 3261 MDL No.: MFCD00000200

Synonyms:  Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
  • CAS Number:99-73-0
  • Formula:C₈H₆Br₂O
  • MDL Number:MFCD00000200
  • Molecular Weight:277.94 g/mol
  • Merck Index:14,1427
  • Boiling Pt:310 °C (1013 hPa)
  • Melting Pt:108…112 °C
  • Storage Temperature:Refrigerator
  • EINECS:202-783-6
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.