Catechol violet
Glass bottle for solids, 25 g
Catalog # B23513.14
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 115-41-3 Molecular Formula: C19H14O7S Formula Weight: 386.38 Beilstein Reference: 353938 MDL No.: MFCD00005868
Synonyms: Pyrocatechol Violet
- CAS Number:115-41-3
- Formula:C₁₉H₁₄O₇S
- MDL Number:MFCD00005868
- Molecular Weight:386.38 g/mol
- Storage Temperature:Ambient
- EINECS:204-088-3
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



