Order Entry
Germany
Orders LinkContactUsLinkComponent
Creatinine ≥98%

Creatinine ≥98%

Glass bottle for solids, 50 g
Catalog # B23097.18
Irritant
Creatinine ≥98%
Catalog # B23097.18
CAS Number:  60-27-5
Creatinine ≥98%
Creatinine ≥98%

Glass bottle for solids, 50 g

CAS Number:  60-27-5
Price
94,50 €
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 98% CAS No.: 60-27-5 Molecular Formula: C4H7N3O Formula Weight: 113.12 Odour: Odourless pH: 7.5-8.5 Melting Point: ca 255° dec. Beilstein Reference: 112061 Merck Reference: 14,2569 MDL No.: MFCD00059730

Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • MDL Number:MFCD00059730
  • Molecular Weight:113.12 g/mol
  • Merck Index:14,2569
  • Melting Pt:∼ 255 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-466-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.