Order Entry
Germany
Orders LinkContactUsLinkComponent
3-Fluoropyrocatechol ≥99%

3-Fluoropyrocatechol ≥99%

Glass bottle for solids, 5 g
Catalog # B22145.06
Corrosive
Irritant
3-Fluoropyrocatechol ≥99%
Catalog # B22145.06
CAS Number:  363-52-0
3-Fluoropyrocatechol ≥99%
3-Fluoropyrocatechol ≥99%

Glass bottle for solids, 5 g

CAS Number:  363-52-0
Price
133,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 363-52-0 Molecular Formula: C6H5FO2 Formula Weight: 128.10 Melting Point: 71-73° Beilstein Reference: 2436074 UN No.: 1759 MDL No.: MFCD00042582

Synonyms:  3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
  • CAS Number:363-52-0
  • Formula:C₆H₅FO₂
  • MDL Number:MFCD00042582
  • Molecular Weight:128.1 g/mol
  • Boiling Pt:218 °C (1013 hPa)
  • Melting Pt:71…73 °C
  • Density:1.514 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.