3-Fluoropyrocatechol ≥99%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
3-Fluoropyrocatechol ≥99%
Catalog # B22145.03
Supplier: Thermo Fisher Scientific
CAS Number: 363-52-0
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 363-52-0 Molecular Formula: C6H5FO2 Formula Weight: 128.10 Melting Point: 71-73° Beilstein Reference: 2436074 UN No.: 1759 MDL No.: MFCD00042582
Synonyms: 3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
- CAS Number:363-52-0
- Formula:C₆H₅FO₂
- MDL Number:MFCD00042582
- Molecular Weight:128.1 g/mol
- Boiling Pt:218 °C (1013 hPa)
- Melting Pt:71…73 °C
- Density:1.514 g/cm³ (20 °C)
- Storage Temperature:Ambient
- ADR:8,III
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



