Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Pentylcyclohexanone ≥98%

4-Pentylcyclohexanone ≥98%

Glass bottle for solids, 50 g
Catalog # B22088.18
4-Pentylcyclohexanone ≥98%
Catalog # B22088.18
CAS Number:  61203-83-6
4-Pentylcyclohexanone ≥98%
4-Pentylcyclohexanone ≥98%

Glass bottle for solids, 50 g

CAS Number:  61203-83-6
Price
186,00 €
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 98+% CAS No.: 61203-83-6 Molecular Formula: C11H20O Formula Weight: 168.28 UN No.: 3082 MDL No.: MFCD00059581

  • CAS Number:61203-83-6
  • Formula:C₁₁H₂₀O
  • MDL Number:MFCD00059581
  • Molecular Weight:168.28 g/mol
  • Density:0.89
  • Storage Temperature:Ambient
  • EINECS:406-670-4
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.