Order Entry
Germany
Orders LinkContactUsLinkComponent
Digoxin 96%

Digoxin 96%

Glass bottle for solids, 1 g
Catalog # B21902.03
Toxic
Digoxin 96%
Catalog # B21902.03
CAS Number:  20830-75-5
Digoxin 96%
Digoxin 96%

Glass bottle for solids, 1 g

CAS Number:  20830-75-5
Price
116,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 96% CAS No.: 20830-75-5 Molecular Formula: C41H64O14 Formula Weight: 780.95 Melting Point: ca 250° dec. Merck Reference: 14,3167 MDL No.: MFCD00003674

  • CAS Number:20830-75-5
  • Formula:C₄₁H₆₄O₁₄
  • MDL Number:MFCD00003674
  • Molecular Weight:780.95 g/mol
  • Merck Index:14,3167
  • Melting Pt:∼ 250 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:244-068-1
  • UN:2811
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.