Order Entry
Germany
Orders LinkContactUsLinkComponent
Cyclooctanone ≥98%

Cyclooctanone ≥98%

Glass bottle for solids, 100 g
Catalog # B20911.22
Corrosive
Cyclooctanone ≥98%
Catalog # B20911.22
CAS Number:  502-49-8
Cyclooctanone ≥98%
Cyclooctanone ≥98%

Glass bottle for solids, 100 g

CAS Number:  502-49-8
Price
87,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 502-49-8 Molecular Formula: C8H14O Formula Weight: 126.20 Flash Point: 72°(161°F) Odour: Pleasant Boiling Point: 195-197° Beilstein Reference: 1280738 UN No.: 1759 MDL No.: MFCD00001754

  • CAS Number:502-49-8
  • Formula:C₈H₁₄O
  • MDL Number:MFCD00001754
  • Molecular Weight:126.2 g/mol
  • Boiling Pt:195…197 °C
  • Melting Pt:37…44 °C
  • Density:0.960
  • Flash Pt:72 °C (161 °F)
  • Storage Temperature:Ambient
  • EINECS:207-940-2
  • UN:1759
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.