Orders LinkContactUsLinkComponent
Cyclooctanone ≥98%
Glass bottle for solids, 25 g
  B20911.14
Cyclooctanone ≥98%
  B20911.14
 :  502-49-8
Cyclooctanone ≥98%
Cyclooctanone ≥98%
Glass bottle for solids, 25 g
 :  502-49-8
32,60 €
1 * 25 g
Purity: 98% CAS No.: 502-49-8 Molecular Formula: C8H14O Formula Weight: 126.20 Flash Point: 72°(161°F) Odour: Pleasant Boiling Point: 195-197° Beilstein Reference: 1280738 UN No.: 1759 MDL No.: MFCD00001754
  •  :502-49-8
  •  :C₈H₁₄O
  •  :MFCD00001754
  •  :126.2 g/mol
  •  :195…197 °C
  •  :37…44 °C
  •  :0.960
  •  :72 °C (161 °F)
  •  :Ambient
  •  :207-940-2
  •  :1759
  •  :8,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g