Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Hydroxybenzhydrazide ≥98%

4-Hydroxybenzhydrazide ≥98%

Glass bottle for solids, 500 g
Catalog # A12702.36
Irritant
4-Hydroxybenzhydrazide ≥98%
Catalog # A12702.36
CAS Number:  5351-23-5
4-Hydroxybenzhydrazide ≥98%
4-Hydroxybenzhydrazide ≥98%

Glass bottle for solids, 500 g

CAS Number:  5351-23-5
Price
474,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98% CAS No.: 5351-23-5 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Melting Point: ca 267° dec. Beilstein Reference: 473052

Synonyms:  4-Hydroxybenzoic hydrazide, 4-Hydroxybenzohydrazide, p-Hydroxybenzoic acid hydrazide, 4-Hydroxybenzene-1-carbohydrazide, 4-Hydroxybenzoic acid hydrazide
  • CAS Number:5351-23-5
  • Formula:C₇H₈N₂O₂
  • MDL Number:MFCD00007605
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:370 °C (1013 hPa)
  • Melting Pt:∼ 267 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:226-326-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications