Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Hydroxybenzhydrazide ≥98%

4-Hydroxybenzhydrazide ≥98%

Glass bottle for solids, 25 g
Catalog # A12702.14
Irritant
4-Hydroxybenzhydrazide ≥98%
Catalog # A12702.14
CAS Number:  5351-23-5
4-Hydroxybenzhydrazide ≥98%
4-Hydroxybenzhydrazide ≥98%

Glass bottle for solids, 25 g

CAS Number:  5351-23-5
Price
41,70 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 5351-23-5 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Melting Point: ca 267° dec. Beilstein Reference: 473052

Synonyms:  4-Hydroxybenzoic hydrazide, 4-Hydroxybenzohydrazide, p-Hydroxybenzoic acid hydrazide, 4-Hydroxybenzene-1-carbohydrazide, 4-Hydroxybenzoic acid hydrazide
  • CAS Number:5351-23-5
  • Formula:C₇H₈N₂O₂
  • MDL Number:MFCD00007605
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:370 °C (1013 hPa)
  • Melting Pt:∼ 267 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:226-326-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.