Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Sodium nitrate ≥98%

Glass bottle for solids, 10.000 g
Catalog # A12327.0C
Oxidizing
Irritant
Sodium nitrate ≥98%
Catalog # A12327.0C
CAS Number:  7631-99-4
Sodium nitrate ≥98%
Sodium nitrate ≥98%

Glass bottle for solids, 10.000 g

CAS Number:  7631-99-4
Price
260,00 €
Log in for availability
1 * 10.000 g
Product Details & Documents

Purity: 98+% CAS No.: 7631-99-4 Molecular Formula: NaNO3 Formula Weight: 84.99 Odour: Odourless Melting Point: ca 308° UN No.: 1498 MDL No.: MFCD00011119

  • CAS Number:7631-99-4
  • Formula:NaNO₃
  • MDL Number:MFCD00011119
  • Molecular Weight:84.99 g/mol
  • Merck Index:14,8647
  • Boiling Pt:380 °C (1013 hPa)
  • Melting Pt:∼308 °C
  • Density:2.26
  • Storage Temperature:Ambient
  • EINECS:231-554-3
  • UN:1498
  • ADR:5.1,III
  • REACH:01-2119488221-41
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10.000 g

Specifications