Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Methylmorpholine ≥99%

4-Methylmorpholine ≥99%

Glass bottle, 500 mL
Catalog # A12158.AP
Flammable
Corrosive
Irritant
4-Methylmorpholine ≥99%
Catalog # A12158.AP
CAS Number:  109-02-4
4-Methylmorpholine ≥99%
4-Methylmorpholine ≥99%

Glass bottle, 500 mL

CAS Number:  109-02-4
Price
52,40 €
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 99% CAS No.: 109-02-4 Molecular Formula: C5H11NO Formula Weight: 101.15 Flash Point: 14°(57°F) Physical Form: Liquid Density: 0.920 Refractive Index: 1.4355 Appearance: Colourless Odour: Amine-like pH: 10.6 Melting Point: -65° Boiling Point: 116-118° Beilstein Reference: 102719 Merck Reference: 14,6277 UN No.: 2535 MDL No.: MFCD00006175

Synonyms:  NMM, N-Methylmorpholine
  • CAS Number:109-02-4
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD00006175
  • Molecular Weight:101.15 g/mol
  • Merck Index:14,6277
  • Boiling Pt:116…118 °C
  • Melting Pt:–65 °C
  • Density:0.920
  • Flash Pt:14 °C (57 °F)
  • Storage Temperature:Ambient
  • EINECS:203-640-0
  • UN:2535
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.