Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4-Methylmorpholine ≥99%

Glass bottle, 100 mL
Catalog # A12158.AE
Flammable
Corrosive
Irritant
4-Methylmorpholine ≥99%
Catalog # A12158.AE
CAS Number:  109-02-4
4-Methylmorpholine ≥99%
4-Methylmorpholine ≥99%

Glass bottle, 100 mL

CAS Number:  109-02-4
Price
25,80 €
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 99% CAS No.: 109-02-4 Molecular Formula: C5H11NO Formula Weight: 101.15 Flash Point: 14°(57°F) Physical Form: Liquid Density: 0.920 Refractive Index: 1.4355 Appearance: Colourless Odour: Amine-like pH: 10.6 Melting Point: -65° Boiling Point: 116-118° Beilstein Reference: 102719 Merck Reference: 14,6277 UN No.: 2535 MDL No.: MFCD00006175

Synonyms:  NMM, N-Methylmorpholine
  • CAS Number:109-02-4
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD00006175
  • Molecular Weight:101.15 g/mol
  • Merck Index:14,6277
  • Boiling Pt:116…118 °C
  • Melting Pt:–65 °C
  • Density:0.920
  • Flash Pt:14 °C (57 °F)
  • Storage Temperature:Ambient
  • EINECS:203-640-0
  • UN:2535
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications