Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyridine ≥99%

Pyridine ≥99%

Glass bottle, 10.000 mL
Catalog # A12005.0D
Flammable
Irritant
Pyridine ≥99%
Catalog # A12005.0D
CAS Number:  110-86-1
Pyridine ≥99%
Pyridine ≥99%

Glass bottle, 10.000 mL

CAS Number:  110-86-1
Price
641,00 €
Log in for availability
1 * 10.000 mL
Product Details & Documents

Purity: 99+% CAS No.: 110-86-1 Molecular Formula: C5H5N Formula Weight: 79.10 Flash Point: 20°(68°F) Physical Form: Liquid Density: 0.978 Refractive Index: 1.5100 Odour: Unpleasant Melting Point: -42° Boiling Point: 115° Solubility: Miscible with water, alcohol, ether, pet ether, oils Beilstein Reference: 103233 Merck Reference: 14,7970 UN No.: 1282 MDL No.: MFCD00011732

Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Merck Index:14,7970
  • Boiling Pt:115 °C
  • Melting Pt:–42 °C
  • Density:0.978
  • Flash Pt:20 °C (68 °F)
  • Storage Temperature:Ambient
  • EINECS:203-809-9
  • UN:1282
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10.000 mL

Specifications