Auftragserfassung
Germany
AuftragsverfolgungKontakt
 

4-Fluorphenol ≥99%

Glasflasche für Feststoffe, 100 g
Catalog # A11945.22
Corrosive
Irritant
4-Fluorphenol ≥99%
Catalog # A11945.22
CAS Number:  371-41-5
4-Fluorphenol ≥99%
4-Fluorphenol ≥99%

Glasflasche für Feststoffe, 100 g

CAS Number:  371-41-5
Price
100,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 371-41-5 Molecular Formula: C6H5FO Formula Weight: 112.10 Flash Point: 68°(154°F) Odour: Phenol-like Boiling Point: 185-188° Beilstein Reference: 1362752 UN No.: 2923 MDL No.: MFCD00002316

Synonyms:  1-Fluor-4-hydroxybenzol
  • CAS Number:371-41-5
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002316
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:185…188 °C
  • Melting Pt:44…50 °C
  • Density:1,22
  • Flash Pt:68 °C (154 °F)
  • Storage Temperature:Raumtemperatur
  • EINECS:206-736-0
  • UN:2923
  • ADR:8,III
Specifications

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.