Order Entry
Germany
Orders LinkContactUsLinkComponent
2'-Methoxyacetophenone ≥98%

2'-Methoxyacetophenone ≥98%

Glass bottle for solids, 100 g
Artikelnummer A11929.22
2'-Methoxyacetophenone ≥98%
Artikelnummer A11929.22
CAS-Nummer:  579-74-8
2'-Methoxyacetophenone ≥98%
2'-Methoxyacetophenone ≥98%

Glass bottle for solids, 100 g

CAS-Nummer:  579-74-8
Preis
144,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 579-74-8 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 108°(226°F) Refractive Index: 1.5393 Boiling Point: 119-120°/10mm Beilstein Reference: 1100602 MDL No.: MFCD00008725

Synonyms:  1-(2-Methoxyphenyl)ethanone
  • CAS Number:579-74-8
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00008725
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:119…120 °C (10 mmHg)
  • Density:1.090
  • Flash Pt:108 °C (226 °F)
  • Storage Temperature:Ambient
  • EINECS:209-446-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications