Order Entry
Germany
Orders LinkContactUsLinkComponent
2'-Methoxyacetophenone ≥98%

2'-Methoxyacetophenone ≥98%

Glass bottle for solids, 25 g
Catalog # A11929.14
2'-Methoxyacetophenone ≥98%
Catalog # A11929.14
CAS Number:  579-74-8
2'-Methoxyacetophenone ≥98%
2'-Methoxyacetophenone ≥98%

Glass bottle for solids, 25 g

CAS Number:  579-74-8
Price
46,80 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 579-74-8 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 108°(226°F) Refractive Index: 1.5393 Boiling Point: 119-120°/10mm Beilstein Reference: 1100602 MDL No.: MFCD00008725

Synonyms:  1-(2-Methoxyphenyl)ethanone
  • CAS Number:579-74-8
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00008725
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:119…120 °C (10 mmHg)
  • Density:1.090
  • Flash Pt:108 °C (226 °F)
  • Storage Temperature:Ambient
  • EINECS:209-446-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.